Learn More
Biocare Medical, LLC TBS Automation Wash Buffer, 20X
SDP
Catalog No. 5083296
Encompass_Preferred
TBS Automation Wash Buffer (sodium azide and thimerosal free) is intended for use in manual and automated immunohistochemistry applications requiring a high quality TBS buffer with superior pH stability. The buffer contains a surfactant that allows reagents to spread uniformly across tissue sections on slides.
Catalog No. 50-832-96 Supplier Biocare Medical, LLC Supplier No. TWB945M
TBS Automation Wash Buffer (sodium azide and thimerosal free) is intended for use in manual and automated immunohistochemistry applications requiring a high quality TBS buffer with superior pH stability. The buffer contains a surfactant that allows reagents to spread uniformly across tissue sections on slides.