Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TBS Buffer

Catalog No. R017R0000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R017R0000 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R017R0000 Supplier Thermo Scientific™ Supplier No. R017R.0000
Add to Cart
Add to Cart

Tris Buffered Saline (TBS) Buffer is supplied as a lyophilized powder and is intended for use with PanPath products.

Tris Buffered Saline (TBS) Buffer is intended for use with PanPath products. TBS is supplied as a lyophilized powder, and when dissolved in one liter distilled or deionised water, it becomes 0.05M TBS, pH 8.0 (0.238M NaCl; 0.0027M KCl) with 0.05% Tween 20.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBS Buffer
For Use With (Equipment) PanPath Products
Physical Form Powder
Product Line REMBRANDT
Quantity 1 Pack
Solution Type Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.