Learn More
Surmodics Inc Tris Buffered Saline/Casein Block & Diluent -10X, 1L
SDP
Catalog No. NC1492474
Encompass_Preferred
BioFX™ Tris Buffered Saline (TBS)/Casein Block and Diluent – 10X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.
Catalog No. NC1492474 Supplier Surmodics Inc Supplier No. TBSC100001
BioFX™ Tris Buffered Saline (TBS)/Casein Block and Diluent – 10X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.