Learn More
Cayman Chemical TBS-Corey Lactone Aldehyde, Purity: ≥95%, CAS: 64091-14-1, 1 g
SDP
Catalog No. 502894633
Encompass_Preferred
TBS-Corey lactone aldehyde is a versatile, hydroxyl-protected intermediate for the synthesis of prostaglandins and prostaglandin analogs. Prostaglandin derivatives with modifications in the alpha chain, the omega chain, or both, are readily accessible from TBS-Corey aldehyde.{5004}
Catalog No. 50-289-4633 Supplier Cayman Chemical Supplier No. 700311 G
TBS-Corey lactone aldehyde is a versatile, hydroxyl-protected intermediate for the synthesis of prostaglandins and prostaglandin analogs. Prostaglandin derivatives with modifications in the alpha chain, the omega chain, or both, are readily accessible from TBS-Corey aldehyde.{5004}