Learn More
Biotang Inc TBS+EDTA (5X), 1000 ML,TBS 5X, EDTA 50 mM, pH 7.4 +/- 0.15. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped and stored at RT. BM-684
Catalog No. 50155735 Shop All BIOTANG Inc Products
Encompass_Preferred
TBS+EDTA (5X), 1000 ML,TBS 5X, EDTA 50 mM, pH 7.4 +/- 0.15. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped and stored at RT. BM-684.
Catalog No. 50-155-735 Supplier Biotang Inc Supplier No. BTBB185
TBS+EDTA (5X), 1000 ML,TBS 5X, EDTA 50 mM, pH 7.4 +/- 0.15. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped and stored at RT. BM-684.