Learn More
Bioworld TBS/Fish Gelatin (10X), 200 mL
Catalog No. 50199167
Encompass_Preferred
TBS fish gelatin solution used as blocking, diluents and coating solutions is supplied as 10X concentrate of 5% solution.Technical Information:5% Fish gelatin in TBS bufferpH range: 7.6 +/- 0.1Sold as 2 x 100mL
Catalog No. 50-199-167 Supplier Bioworld Supplier No. 217610581
TBS fish gelatin solution used as blocking, diluents and coating solutions is supplied as 10X concentrate of 5% solution.Technical Information:5% Fish gelatin in TBS bufferpH range: 7.6 +/- 0.1Sold as 2 x 100mL