Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc TBS HCl (10X), pH 7.2, 500 mL
SDP

Catalog No. 507517377
Encompass_Preferred

Tris HCl (10X), pH 7.2 is a Buffered Saline Solution used in procedures for the isolation of DNA.

Filtered through a 0.22 µm membrane
DNase, RNase, and Protease tested

Applications: Tris buffered saline is useful for the following: as a biological diluent buffer, as a buffer for antibodies, as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health

Catalog No. 50-751-7377 Supplier Quality Biological Inc Supplier No. 351058101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tris HCl (10X), pH 7.2 is a Buffered Saline Solution used in procedures for the isolation of DNA.

Filtered through a 0.22 µm membrane
DNase, RNase, and Protease tested

Applications: Tris buffered saline is useful for the following: as a biological diluent buffer, as a buffer for antibodies, as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.