Learn More
General Data Healthcare™ Knife Holder for CUT™ Rotary Microtomes
SDP
Catalog No. 22046827 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Product Type:
Knife Holder
Knife Holder Base
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-046-827 Knife Holder Base
22-046-829 Knife Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-046-827 Supplier General Data Healthcare™ Supplier No. CUT2000
  • Knife holder for conventional steel or tungsten carbide-edged knives or disposable blade holders
  • Knife holder must be used with Cat. No. 22-046-827 universal knife holder base
Specifications
Product Type Knife Holder Base
For Use With (Equipment) Cut Rotary Microtome