Learn More
General Data Healthcare™ Knife Holder for CUT™ Rotary Microtomes
Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Description
- Knife holder for conventional steel or tungsten carbide-edged knives or disposable blade holders
- Knife holder must be used with Cat. No. 22-046-827 universal knife holder base
Specifications
Specifications
| Product Type | Knife Holder Base |
| For Use With (Equipment) | Cut Rotary Microtome |