Learn More
General Data Healthcare™ Knives for CUT™ Rotary Microtomes
SDP
Catalog No. 22046853 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Product Type:
Tungsten Carbide-edged Knife
Catalog No. Product Type
22-046-853 Tungsten Carbide-edged Knife
1 options
Catalog No. 22-046-853 Supplier General Data Healthcare™ Supplier No. CUT3020
Specifications
Product Type Tungsten Carbide-edged Knife
Profile D
For Use With (Equipment) Cut Rotary Microtome
Material Tungsten Carbide
Length (English) 6.2 in.
Length (Metric) 16 cm