Learn More
Quality Biological Inc TBS 10X pH 7.4, 1000 mL
SDP
Catalog No. 507517351
Encompass_Preferred
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques. ¿ DNase, RNase and Protease tested. ¿ Applications: TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase, and Protease Tested ¿ pH: 7.4¿0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7351 Supplier Quality Biological Inc Supplier No. 351086131
Add to Cart
Add to Cart
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques. ¿ DNase, RNase and Protease tested. ¿ Applications: TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase, and Protease Tested ¿ pH: 7.4¿0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months