Learn More
General Data Healthcare™ SHUR/Mark™ Coverglasses
SDP
Catalog No. 22046402 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Size: 24 x 50mm
Catalog No.
22-046-402 Size: 24 x 50mm
1 options
This item is not returnable. View return policy
Catalog No. 22-046-402 Supplier General Data Healthcare™ Supplier No. SM2450
This item is not returnable. View return policy
Ideal for use in automated coverslipping instruments
SHUR/Mark™ premium coverglass is made from pure white flat sheets of hydrolytic Class l borosilicate glass (refractive index ne=1.525±).
  • Ultrasonically precleaned
  • Packaged in plastic boxes with moisture-absorbing desiccant
  • Hermetically sealed outer packaging for longer shelf life
  • No self-fluorescence
  • Ideal for humid environments
  • Thickness No. 1
Compliance
Manufactured in accordance with Din ISO 8255-1:2003-05.