Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TBS Tablets, Calbiochem™,

Catalog No. 5247501EA Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Strip
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-475-01EA 1 Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-475-01EA Supplier MilliporeSigma™ Supplier No. 5247501EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dissolving one tablet in 500mL of deionized H2O yields 150mM NaCl, 50mM Tris-HCl buffer

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBS Tablets
pH 7.6
Physical Form Tablet
Quantity 1 Strip
Solution Type Buffer
Synonym Tris Buffered Saline 10 Tablets
Solubility Information Soluble in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.