Learn More
Aniara Diagnostica LLC TBS TBLT PH 7.6 500ML 100 EA
Catalog No. 501008790
Encompass
Medicago Tris Buffered Saline 1x (TBS) pH 7.6 supplied as preweighed tablets yields 500 mL 0.050 Tris-HCl, 0.15 M KCl with pH 7.6 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.
Catalog No. 50-100-8790 Supplier Aniara Diagnostica LLC Supplier No. A097500100
Medicago Tris Buffered Saline 1x (TBS) pH 7.6 supplied as preweighed tablets yields 500 mL 0.050 Tris-HCl, 0.15 M KCl with pH 7.6 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.