Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ TMD™-7 Tissue Marking Dyes Refills
SDP

Catalog No. 1518344 Shop All Triangle Biomedical Sciences Inc. (TBS) Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-183-44 Blue
15-183-45 Green
15-183-43 Black
15-183-46 Red
15-183-47 Yellow
22-047-154 Purple
15-183-23 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 15-183-44 Supplier General Data Healthcare™ Supplier No. TMDBL
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refills for the TMD-7 Tissue Marking Dyes 7 Color Kit

Specifications

Capacity 8 oz.
Color Blue
Product Type Tissue Marking Dye
Quantity 4 Case
Solution Type Stain
Formulation Ammonium Hydroxide 2%, Copper Phthalocyanine 17%, Formaldehyde 0.05%, Water 80%
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.