Learn More
Biotang Inc TBS/TWEEN-20 (10X) 500 ML, IBB-580, 1% Tween-20 in 250 mM Tris-HCl, 27 mM KCl and 1.37 M NaCl, pH 7.4 +/- 0.2. Prepared in 18.2 megohm water.and autoclaved at 15 psi for 30 min on liquid cycle.
Catalog No. 50152234 Shop All BIOTANG Inc Products
Encompass_Preferred
TBS/TWEEN-20 (10X) 500 ML, IBB-580, 1% Tween-20 in 250 mM Tris-HCl, 27 mM KCl and 1.37 M NaCl, pH 7.4 +/- 0.2. Prepared in 18.2 megohm water.and autoclaved at 15 psi for 30 min on liquid cycle.
Catalog No. 50-152-234 Supplier Biotang Inc Supplier No. BTBB781
TBS/TWEEN-20 (10X) 500 ML, IBB-580, 1% Tween-20 in 250 mM Tris-HCl, 27 mM KCl and 1.37 M NaCl, pH 7.4 +/- 0.2. Prepared in 18.2 megohm water.and autoclaved at 15 psi for 30 min on liquid cycle.