Learn More
Research Products International Corp TBS / Tween 20 10X Solution, 500 Milliliters
SDP
Catalog No. 50712771
Encompass_Preferred
Formulation contains 0.5% Tween 20 in 10X TB.
Catalog No. 50-712-771 Supplier Research Products International Corp Supplier No. T26100500.0
Formulation contains 0.5% Tween 20 in 10X TB.