Learn More
Alkali Scientific TBS / Tween 20 10X Solution, 500 Milliliters
SDP
Catalog No. 5000661018
Encompass_Preferred
TBS / Tween 20 10X Solution is a highly effective washing buffer used to minimize background noise during immunodetection procedures such as Western blotting and ELISA. This 500mL solution is ideal for smaller-scale applications or as a concentrate that can be diluted for use in various experimental setups. The Tris component provides pH stability, while Tween 20 aids in reducing non-specific binding during protein-protein interactions. This formulation ensures cleaner results by washing away excess proteins and antibodies that do not bind specifically to the target. TBS-Tween 20 is essential for improving the signal-to-noise ratio in sensitive immunoassays and diagnostic tests.
Catalog No. 50-006-61018 Supplier Alkali Scientific Supplier No. TBS002CP
TBS / Tween 20 10X Solution is a highly effective washing buffer used to minimize background noise during immunodetection procedures such as Western blotting and ELISA. This 500mL solution is ideal for smaller-scale applications or as a concentrate that can be diluted for use in various experimental setups. The Tris component provides pH stability, while Tween 20 aids in reducing non-specific binding during protein-protein interactions. This formulation ensures cleaner results by washing away excess proteins and antibodies that do not bind specifically to the target. TBS-Tween 20 is essential for improving the signal-to-noise ratio in sensitive immunoassays and diagnostic tests.