Learn More
Boston Bioproducts Inc Tris Buffered Saline-Tween (TBST), 20X Concentration, 4 x 1 L, 2% Tween-20, pH 7.40 ± 0.20, Wash buffer for immunoassays, Antibody diluent
Catalog No. NC2145143
Encompass
Tris Buffered Saline-Tween (TBST), 20X Concentration, 4 x 1 L, 2% Tween-20, pH 7.40 ± 0.20, Wash buffer for immunoassays, Antibody diluent
This item is not returnable. View return policy
Catalog No. NC2145143 Supplier Boston Bioproducts Inc Supplier No. IBB180X4X1L
Add to Cart
Add to Cart
Tris Buffered Saline-Tween (TBST), 20X Concentration, 4 x 1 L, 2% Tween-20, pH 7.40 ± 0.20, Wash buffer for immunoassays, Antibody diluent