Learn More
Boston Bioproducts Inc Tris Buffered Saline-Tween, TBST (20X, with 2% Tween-20, pH 7.4) - 1L
Catalog No. NC2145142
Encompass
20X Tris Buffered Saline - Tween (TBST) is a buffer-surfactant solution supporting a stable pH range between 7.4 and 8.0. When combined in a variable ratio, TBST is an effective wash buffer in Western blotting, in situ hybridization procedures, and immunohistochemistry staining, as it minimizes non specific binding of the materials without disrupting antigen-antibody binding. TBST is also used as an antibody diluent in immunoassays, and supports an array of molecular biology techniques.
This item is not returnable. View return policy
Catalog No. NC2145142 Supplier Boston Bioproducts Inc Supplier No. IBB180X1L
20X Tris Buffered Saline - Tween (TBST) is a buffer-surfactant solution supporting a stable pH range between 7.4 and 8.0. When combined in a variable ratio, TBST is an effective wash buffer in Western blotting, in situ hybridization procedures, and immunohistochemistry staining, as it minimizes non specific binding of the materials without disrupting antigen-antibody binding. TBST is also used as an antibody diluent in immunoassays, and supports an array of molecular biology techniques.