Learn More
Alkali Scientific TBS with Tween, 20X, 7.4 pH, Sterile Buffered, 1 Liter
SDP
Catalog No. 5000660240
Encompass_Preferred
TBS with Tween 20X, pH 7.4, is a sterile buffered solution commonly used in biochemical and molecular biology applications. This 1-liter bottle provides a high-concentration stock solution that can be diluted to a working concentration for various experimental protocols, including protein assays, immunohistochemistry, and Western blotting. The inclusion of Tween 20 as a detergent helps minimize non-specific binding in protein detection and other assays, ensuring clearer, more reliable results. The sterile, buffered formulation maintains a stable pH of 7.4, providing optimal conditions for biological experiments that require precise buffering and detergent inclusion to reduce interference.
Catalog No. 50-006-60240 Supplier Alkali Scientific Supplier No. BSW001CP
TBS with Tween 20X, pH 7.4, is a sterile buffered solution commonly used in biochemical and molecular biology applications. This 1-liter bottle provides a high-concentration stock solution that can be diluted to a working concentration for various experimental protocols, including protein assays, immunohistochemistry, and Western blotting. The inclusion of Tween 20 as a detergent helps minimize non-specific binding in protein detection and other assays, ensuring clearer, more reliable results. The sterile, buffered formulation maintains a stable pH of 7.4, providing optimal conditions for biological experiments that require precise buffering and detergent inclusion to reduce interference.