Learn More
Biotang Inc TBS WITH TRITON (10X), 500 ML, BM-300T, TBS 10X, Triton X 100 1%. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped and stored at RT.
Catalog No. 50155731 Shop All BIOTANG Inc Products
Encompass_Preferred
TBS WITH TRITON (10X), 500 ML, BM-300T, TBS 10X, Triton X 100 1%. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped and stored at RT.
Catalog No. 50-155-731 Supplier Biotang Inc Supplier No. BTBB181
TBS WITH TRITON (10X), 500 ML, BM-300T, TBS 10X, Triton X 100 1%. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped and stored at RT.