Learn More
Research Products International Corp TBS with Tween, 20X, 7.4 pH, Sterile Buffered, 1 Liter
SDP
Catalog No. 501368661
Encompass_Preferred
TBS with Tween, 20X, 7.4 pH, Sterile Buffered is a commonly used buffer solution in the life science marketplace. TBS stands for Tris-Buffered Saline, and Tween refers to Tween 20, a nonionic detergent.
TBS with Tween is primarily used in various applications such as Western blotting, immunohistochemistry, and antibody-based assays. It serves as a buffer for sample dilution, antibody washing, and blocking steps during these experimental procedures.
The buffer solution is prepared at a 20X concentration, meaning it is twenty times more concentrated than the final desired concentration. This concentrated form allows for efficient storage and transportation.
TBS with Tween, 20X, 7.4 pH, Sterile Buffered is specifically formulated to have a pH of 7.4, which is the physiological pH of many biological systems.
Catalog No. 50-136-8661 Supplier Research Products International Corp Supplier No. T591801000.0
TBS with Tween, 20X, 7.4 pH, Sterile Buffered is a commonly used buffer solution in the life science marketplace. TBS stands for Tris-Buffered Saline, and Tween refers to Tween 20, a nonionic detergent.
TBS with Tween is primarily used in various applications such as Western blotting, immunohistochemistry, and antibody-based assays. It serves as a buffer for sample dilution, antibody washing, and blocking steps during these experimental procedures.
The buffer solution is prepared at a 20X concentration, meaning it is twenty times more concentrated than the final desired concentration. This concentrated form allows for efficient storage and transportation.
TBS with Tween, 20X, 7.4 pH, Sterile Buffered is specifically formulated to have a pH of 7.4, which is the physiological pH of many biological systems.