Learn More
Cayman Chemical TBTA, 510758-28-8, 500 mg
SDP
Supplier:  Cayman Chemical 18816500
Encompass_Preferred
Molecular Formula C30H30N10, Purity ≥98%, Formula Weight 530.6, Synonyms: Tris(benzyltriazolylmethyl)amine; A tertiary amine that complexes with, and stabilizes, Cu(I) to accelerate azide-alkyne cycloadditions, as used in click chemistry.
Catalog No. NC1355341