Learn More
AnaSpec TBTA,TRIS(1-BNZYL-1H-1,2(50MG)
Catalog No. 50352436
Encompass
TBTA, Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl] amine, ; MW 530.6; (50 mg) A ligand used in the synthesis of a Cu(I) catalyst for the synthesis of N-sulfonylazetidin-2-imines.1,2
Catalog No. 50352436 Supplier AnaSpec Supplier No. AS6336050
TBTA, Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl] amine, ; MW 530.6; (50 mg) A ligand used in the synthesis of a Cu(I) catalyst for the synthesis of N-sulfonylazetidin-2-imines.1,2