Learn More
Strem, An Ascensus Company CAS# [511292-99-2]. 1g. (t-Butylimido)tris(ethylmethylamino)tantalum(V), (99.99%-Ta) PURATREM TBTEMT.
SDP
Supplier:  Strem, An Ascensus Company 7307351G
Encompass
CAS# 511292-99-2. 1g. (t-Butylimido)tris(ethylmethylamino)tantalum(V), (99.99%-Ta) PURATREM TBTEMT. . Molecular Weight: 426.38. Molecular Formula: C13H33N4Ta. Color/form: yellow liq. Strem# 73-0735. www.strem.com/catalog/v3/73-0735/
Catalog No. 50-197-9356