Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tbubrettphos Pd G3

Catalog No. 7459795G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-597-95G 5 g
74-597-9100MG 100 mg
74-597-92G 2 g
74-597-9500MG 500 mg
74-597-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 74-597-95G Supplier Sigma Aldrich Supplier No. 7459795G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tbubrettphos Pd G3

Specifications

CAS 1536473-72-9
Melting Point 119°C to 131°C
Molecular Formula C44H62NO5PPdS
Linear Formula C44H62NO5PPdS
Quantity 5 g
MDL Number MFCD25976528
Synonym tert-BuBrettPhos-Pd-G3; [(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate
Molecular Weight (g/mol) 854.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.