Learn More
Strem, An Ascensus Company tBuXPhos PalladacycleG3 98% 5g
SDP
Supplier:  Strem, An Ascensus Company 4603235G
Encompass
CAS# 1447963-75-8. 5g. Methanesulfonato(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [t-BuXPhos Palladacycle Gen. 3]. MFCD22666411. Molecular Weight: 794.38. Molecular Formula: C42H58NO3PPdS. Color/form: yellow to light brown Strem# 46-0323. http://www.strem.com/catalog/v/46-0323/
Catalog No. 50-203-4163