Learn More
Aviva Systems Biology Corp TBX1 Antibody - C-terminal region (ARP33399_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33399_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TBX1. Alternate names: DGS, TGA, CAFS, CTHM, DGCR, DORV, VCFS, TBX1CApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52904