Learn More
Aviva Systems Biology Corp TBX1 Antibody - middle region (ARP33398_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBX1. Alternate names: CAFS, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCFSApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52903