Learn More
Aviva Systems Biology Corp TBX15 Antibody - C-terminal region (ARP47723_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47723_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TBX15. Alternate names: TBX14ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53527