Learn More
Aviva Systems Biology Corp TBX15 Antibody - C-terminal region (ARP47725_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47725_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TBX15. Alternate names: TBX14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59653