Learn More
Aviva Systems Biology Corp Tbx18 Antibody - middle region (ARP37915_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tbx18. It was validated on Western Blot. Tbx18,T-box18,2810012F10Rik; 2810404D13Rik; Tbx18,T-box transcription factor TBX18,T-box transcription factor TBX18,T-box protein 18The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810012F10Rik, 2810404D13RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-701