Learn More
Aviva Systems Biology Corp TBX18 Antibody - C-terminal region (ARP37832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TBX18. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56446