Learn More
Aviva Systems Biology Corp TBX19 Antibody - N-terminal region (ARP32363_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32363_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX19. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TBX19. Alternate names: TPIT, TBS19, dJ747L4.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54545