Learn More
Aviva Systems Biology Corp TBX20 Antibody - middle region (ARP40123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBX20. Alternate names: ASD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57300