Learn More
Aviva Systems Biology Corp TBX21 Antibody - C-terminal region (ARP37251_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37251_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse TBX21. Alternate names: TBT1, Tbet, TblymApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53067