Learn More
Aviva Systems Biology Corp TBX21 Antibody - middle region (ARP32612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX21. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TBX21. Alternate names: HGNC:11599, T-PET, T-bet, TBLYM, TBETApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54674