Learn More
Aviva Systems Biology Corp TBX22 Antibody - middle region (ARP32745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBX22. Alternate names: CLPA, TBXX, dJ795G23.1, CPXApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54738