Learn More
Aviva Systems Biology Corp Tbx22 Antibody - N-terminal region (ARP37488_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37488_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tbx22. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: D230020M15RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-642