Learn More
Aviva Systems Biology Corp TBX3 Antibody - middle region (ARP33412_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33412_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBX3. Alternate names: UMS, XHL, TBX3-ISOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55008