Learn More
Aviva Systems Biology Corp TBX5 Antibody - middle region (ARP33402_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33402_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TBX5. Alternate names: HOSApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54998