Learn More
Aviva Systems Biology Corp TBX6 Antibody - N-terminal region (ARP33405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBX6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TBX6. Alternate names: DFNB67ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55002