Learn More
Aviva Systems Biology Corp TBXA2R Antibody - N-terminal region (ARP64719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBXA2R. It was validated on Western Blot. . Alternate names: TXA2-R, BDPLT13ApplicationWB. Species reactivity: Cow, Dog, Human
Catalog No. 50-288-344