Learn More
Eurofins Discoverx ChemiBrite TP Prostanoid Receptor Stable Cell Line, 2 vials. (HTS081L)
Supplier:  Eurofins Discoverx HTS081L
Encompass
The ChemiBrite TP Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin H2, thromboxane A2) based activation of TBXA2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses TBXA2R alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of TBXA2R, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5822
Add to Cart
Request a Quote