Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ TC-100 Insect Medium
GREENER_CHOICE

Catalog No. 13055025
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-055-025 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-055-025 Supplier Gibco™ Supplier No. 13055025
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC-100 Insect Medium

TC 100 Insect Medium was originally designed for the growth of insect cells, including Spodeptera frugiperda (Sf9), and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ Media options for a range of insect cell culture applications. TC 100 Insect Medium is rich in amino acids and uses tryptose as an additional sugar source.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ TC 100 Insect Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Gibco™ TC 100 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. TC 100 Insect Medium requires ambient CO2 in order to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Form Liquid
Product Type Insect Cell Medium
With Additives Glutamine, Tryptose Phosphate Broth, Sodium Bicarbonate
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Insect Cell Culture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.