Learn More
Cayman Chemical TC 14012 (hydrochloride), Purity: ≥95%, 5 mg
SDP
Catalog No. 502915573
Encompass_Preferred
A CXCR4 antagonist (IC50 = 2.9 nM) and CXCR7 agonist; induces β-arrestin recruitment in HEK293T cells expressing CXCR7 (EC50 = 350 nM); reduces the cytopathic effect of HIV in infected MT-4 cells (EC50 = 0.4 nM); inhibits HIV entry in vitro in a CXCR4-dependent manner (IC50 = 19.3 nM); inhibits CXCL12-induced migration in MDA-MB-231 cells and HUVECs from 10-1,000 nM; decreases infarct size in a mouse model of LAD coronary artery ligation-induced acute myocardial infarction at 10 mg/kg
Catalog No. 50-291-5573 Supplier Cayman Chemical Supplier No. 381405 MG
A CXCR4 antagonist (IC50 = 2.9 nM) and CXCR7 agonist; induces β-arrestin recruitment in HEK293T cells expressing CXCR7 (EC50 = 350 nM); reduces the cytopathic effect of HIV in infected MT-4 cells (EC50 = 0.4 nM); inhibits HIV entry in vitro in a CXCR4-dependent manner (IC50 = 19.3 nM); inhibits CXCL12-induced migration in MDA-MB-231 cells and HUVECs from 10-1,000 nM; decreases infarct size in a mouse model of LAD coronary artery ligation-induced acute myocardial infarction at 10 mg/kg