Learn More
Cole-Parmer Techne F937D Temp Controller for Small Fluidized Sand Baths
Supplier:  Cole-Parmer 0118450
Encompass_Preferred
This temperature controller features fast heat-ups and accurate temperature control to 600C. It integrates an air flowmeter and a PID temperature control with dual-setpoint programming to improve accuracy, increase temperature stability and allow temperature settings to 1C/F. It comes with a chromel or alumel type K thermocouple, which fits into the baths thermowell, and 10 feet of 3/8 inch tubing for air flow control.
Catalog No. 50-196-0463