Learn More
Medchemexpress LLC TC-A 2317 hydrochloride | 1245907-03-2 | MFCD19690936 | >=98.0% | 392.93 | C19H29ClN6O | 1 MG
SDP
Catalog No. 5000247538
Encompass_Preferred
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a small-molecule Aurora kinase inhibitor used in cell-cycle and DNA damage research. It is supplied as a research reagent for biochemical and cellular studies.
  • Used in cell-cycle and DNA damage pathway studies.
  • Hydrochloride salt form for improved stability and solubility.
  • Molecular formula C19H29ClN6O; molecular weight 392.93.
  • Available in small pack sizes suitable for screening and assay work.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-47538 Supplier Medchemexpress LLC Supplier No. HY1032661MG
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a small-molecule Aurora kinase inhibitor used in cell-cycle and DNA damage research. It is supplied as a research reagent for biochemical and cellular studies.
  • Used in cell-cycle and DNA damage pathway studies.
  • Hydrochloride salt form for improved stability and solubility.
  • Molecular formula C19H29ClN6O; molecular weight 392.93.
  • Available in small pack sizes suitable for screening and assay work.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.