Learn More
Medchemexpress LLC TC-A 2317 (hydrochloride) | 1245907-03-2 | MFCD19690936 | 99.6% | 392.93 | C19H29ClN6O | 5 MG
SDP
Catalog No. 5000246839
Encompass_Preferred
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a potent, orally active Aurora A kinase inhibitor used for research applications. It demonstrates high potency, selectivity, and antiproliferative activity in cancer cell models.
  • Potent Aurora A inhibitor with Ki = 1.2 nM
  • Selective over Aurora B (Ki = 101 nM)
  • Antiproliferative activity in HCT-116 cells (IC50 = 115 nM)
  • Hydrochloride salt for improved stability and handling
  • High measured purity (~99.6%) suitable for research use
  • Available in small-mass packaging for laboratory studies
Catalog No. 50-002-46839 Supplier Medchemexpress LLC Supplier No. HY1032665MG
TC-A 2317 hydrochloride is the hydrochloride salt of TC-A 2317, a potent, orally active Aurora A kinase inhibitor used for research applications. It demonstrates high potency, selectivity, and antiproliferative activity in cancer cell models.
  • Potent Aurora A inhibitor with Ki = 1.2 nM
  • Selective over Aurora B (Ki = 101 nM)
  • Antiproliferative activity in HCT-116 cells (IC50 = 115 nM)
  • Hydrochloride salt for improved stability and handling
  • High measured purity (~99.6%) suitable for research use
  • Available in small-mass packaging for laboratory studies